Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhodium(III) acetylacetonate

Catalog No. 2827745G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-277-45G 5 g
28-277-4250MG 250 mg
28-277-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-277-45G Supplier Sigma Aldrich Supplier No. 2827745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rhodium(III) acetylacetonate

Specifications

CAS 14284-92-5
Melting Point 263°C to 264°C
Molecular Formula C15H21O6Rh
Linear Formula [CH3COCH=C(O-)CH3]3 Rh
Quantity 5 g
MDL Number MFCD00083144
Synonym 2,4-Pentanedione rhodium(III) derivative; Rh(acac)3
Molecular Weight (g/mol) 400.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.