Learn More
Strem, An Ascensus Company Rhodium(III)chloridehydrate 1g
SDP
Catalog No. 5090112249
Encompass
CAS# 20765-98-4. 1g. Rhodium(III) chloride hydrate (38-41% Rh). MFCD00149839. Molecular Weight: 209.28. Molecular Formula: RhCl3∙XH2O. Color/form: dark red pwdr. Strem# 45-1880. http://www.strem.com/catalog/v/45-1880/
Catalog No. 50-901-12249 Supplier Strem, An Ascensus Company Supplier No. 4518801G
CAS# 20765-98-4. 1g. Rhodium(III) chloride hydrate (38-41% Rh). MFCD00149839. Molecular Weight: 209.28. Molecular Formula: RhCl3∙XH2O. Color/form: dark red pwdr. Strem# 45-1880. http://www.strem.com/catalog/v/45-1880/