Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rhodium(III) oxide | 12036-35-0 | MFCD00011205 | 253.81 g/mol | Rh2O3 | 100 MG
SDP

Catalog No. 5000301776
Encompass_Preferred

Rhodium(III) oxide is an inorganic rhodium oxide reagent (Rh2O3) commonly used in catalyst development, materials research, and other laboratory applications requiring rhodium oxides. The product is provided as a small-scale powder suitable for experimental use, with CAS 12036-35-0 and a molecular weight of 253.81 g/mol.

  • Inorganic rhodium oxide powder for research use.
  • Common applications include catalyst development and materials research.
  • CAS number 12036-35-0 for unambiguous identification.
  • Molecular formula Rh2O3 and molecular weight 253.81 g/mol.
  • Supplied in a 100 mg pack suitable for small-scale experiments.

Catalog No. 50-003-01776 Supplier Medchemexpress LLC Supplier No. HYW033823100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rhodium(III) oxide is an inorganic rhodium oxide reagent (Rh2O3) commonly used in catalyst development, materials research, and other laboratory applications requiring rhodium oxides. The product is provided as a small-scale powder suitable for experimental use, with CAS 12036-35-0 and a molecular weight of 253.81 g/mol.

  • Inorganic rhodium oxide powder for research use.
  • Common applications include catalyst development and materials research.
  • CAS number 12036-35-0 for unambiguous identification.
  • Molecular formula Rh2O3 and molecular weight 253.81 g/mol.
  • Supplied in a 100 mg pack suitable for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.