Learn More
Medchemexpress LLC Rhodium(III) oxide | 12036-35-0 | MFCD00011205 | 253.81 | Rh2O3 | 500 MG
SDP
Catalog No. 5000299415
Encompass_Preferred
Rhodium(III) oxide is an inorganic rhodium oxide used as a heterogeneous catalyst and in materials and energy research, including fuel cell and catalyst development. Supplied as a laboratory powder for research use, users should consult the product datasheet and certificate of analysis for batch-specific purity, handling, and storage information.
  • High catalytic activity for hydrogenation and oxidation reactions.
  • Applicable to fuel cell and energy materials research.
  • Available in small laboratory pack sizes for research applications.
  • Solid powder form suitable for material synthesis and catalyst preparation.
  • Purity and storage conditions provided in the certificate of analysis.
Catalog No. 50-002-99415 Supplier Medchemexpress LLC Supplier No. HYW033823500MG
Rhodium(III) oxide is an inorganic rhodium oxide used as a heterogeneous catalyst and in materials and energy research, including fuel cell and catalyst development. Supplied as a laboratory powder for research use, users should consult the product datasheet and certificate of analysis for batch-specific purity, handling, and storage information.
  • High catalytic activity for hydrogenation and oxidation reactions.
  • Applicable to fuel cell and energy materials research.
  • Available in small laboratory pack sizes for research applications.
  • Solid powder form suitable for material synthesis and catalyst preparation.
  • Purity and storage conditions provided in the certificate of analysis.