Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhodium On Alumina

Catalog No. 2128575G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-285-75G 5 g
21-285-725G 25 g
21-285-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-285-75G Supplier Sigma Aldrich Supplier No. 2128575G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Rhodium on alumina (Rh/Al2O3) (5 wt. % loading, matrix alumina support, powder) is utilized as a catalyst for the steam reforming of ethanol for use in hydrogen production using molten carbonate fuel cells. Rh/Al2O3 can also be used as a catalyst in the production of hydrogen from sodium borohydride (NaBH4).

Specifications

MDL Number MFCD00011201
Quantity 5 g
Synonym Rh/Al2O3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.