Learn More
Sigma Aldrich Rhodium On Carbon
Catalog No. 20616425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-616-425G 25 g
20-616-41G 1 g
20-616-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-616-425G Supplier Sigma Aldrich Supplier No. 20616425G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Rhodium on carbon (Rh/C) can be used as a catalyst in the reductive transformation of carbonyl compounds. It can be used as a heterogeneous catalyst for the production of formaldehyde through a greener technique.
Specifications
Quantity 25 g
MDL Number MFCD00011201