Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies Rhodium Standard, 1,000 μg/mL, 125 mL

Catalog No. NC2258624
Encompass

Agilent™ Rhodium (Rh) standard solution provides reliable and accurate calibrations for ICP-MS analyses. This high-purity single-element reference material is manufactured with NIST traceability to ensure consistent instrument performance. The solution is formulated as a single component mixture of Rhodium (III) chloride hydrate (ICP grade) at a concentration of 1,000 μg/mL in a solvent of water and dilute hydrochloric acid. This standard is supplied in a 125 mL bottle.

  • Chemical type: Inorganic
  • Quality standard: ISO 17034
  • Concentration: 1,000 μg/mL
  • Volume: 125 mL
  • Analysis type: Quantitative
  • CAS number: 10049-07-7

This item is not returnable. View return policy
Catalog No. NC2258624 Supplier Agilent Technologies Supplier No. ICP045
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agilent™ Rhodium (Rh) standard solution provides reliable and accurate calibrations for ICP-MS analyses. This high-purity single-element reference material is manufactured with NIST traceability to ensure consistent instrument performance. The solution is formulated as a single component mixture of Rhodium (III) chloride hydrate (ICP grade) at a concentration of 1,000 μg/mL in a solvent of water and dilute hydrochloric acid. This standard is supplied in a 125 mL bottle.

  • Chemical type: Inorganic
  • Quality standard: ISO 17034
  • Concentration: 1,000 μg/mL
  • Volume: 125 mL
  • Analysis type: Quantitative
  • CAS number: 10049-07-7

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.