Learn More
Medchemexpress LLC Rhodizonic acid diso 25g
SDP
Catalog No. 5000205456
Encompass_Preferred
Rhodizonic acid disodium (Sodium rhodizonate dibasic) can be used as an indicator in the volumetric determination of barium and sulfates Rhodizonic acid disodium is a kind of biological materials or organic compounds that are widely used in life science research[1]
Catalog No. 50-002-05456 Supplier Medchemexpress LLC Supplier No. 25GHYW115721
Rhodizonic acid disodium (Sodium rhodizonate dibasic) can be used as an indicator in the volumetric determination of barium and sulfates Rhodizonic acid disodium is a kind of biological materials or organic compounds that are widely used in life science research[1]