Learn More
Medchemexpress LLC Rhodizonic acid diso 5g
SDP
Catalog No. 5000206662
Encompass_Preferred
Rhodizonic acid disodium (Sodium rhodizonate dibasic) can be used as an indicator in the volumetric determination of barium and sulfates Rhodizonic acid disodium is a kind of biological materials or organic compounds that are widely used in life science research[1]
Catalog No. 50-002-06662 Supplier Medchemexpress LLC Supplier No. HYW1157215G
Rhodizonic acid disodium (Sodium rhodizonate dibasic) can be used as an indicator in the volumetric determination of barium and sulfates Rhodizonic acid disodium is a kind of biological materials or organic compounds that are widely used in life science research[1]