Learn More
ATCC Ureaplasma parvum
Catalog No. 502381203
Encompass
Ureaplasma parvum strain U26 was isolated from a pregnant woman. This strain requires an additional 5-10% horse serum added to the broth for growth.
Catalog No. 50-238-1203 Supplier ATCC Supplier No. 33697
Ureaplasma parvum strain U26 was isolated from a pregnant woman. This strain requires an additional 5-10% horse serum added to the broth for growth.