Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rhodojaponin III | 26342-66-5 | 1 MG
SDP

Catalog No. 5000421730
Encompass_Preferred

Rhodojaponin III is a diterpenoid extracted from the leaves of Rhododendron molle. It exhibits anti-inflammatory activity and is intended for research use only.

  • Diterpenoid compound
  • Derived from Rhododendron molle leaves
  • Demonstrates anti-inflammatory activity
  • Molecular formula: C20H32O6
  • Molecular weight: 368.46
  • Purity: 99.83%
  • Appearance: White to off-white solid
  • Store at 4°C, protected from light
  • For solvent storage, protect from light and store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (≥ 3.68 mg/mL)

Catalog No. 50-004-21730 Supplier Medchemexpress LLC Supplier No. HYN21521MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rhodojaponin III is a diterpenoid extracted from the leaves of Rhododendron molle. It exhibits anti-inflammatory activity and is intended for research use only.

  • Diterpenoid compound
  • Derived from Rhododendron molle leaves
  • Demonstrates anti-inflammatory activity
  • Molecular formula: C20H32O6
  • Molecular weight: 368.46
  • Purity: 99.83%
  • Appearance: White to off-white solid
  • Store at 4°C, protected from light
  • For solvent storage, protect from light and store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (≥ 3.68 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.