Learn More
Medchemexpress LLC Rhodojaponin V | 37720-86-8 | 5 MG
SDP
Catalog No. 5000418351
Encompass_Preferred
Rhodojaponin V is a diterpenoid isolated from the leaves of *Rhododendron molle* and is recognized for its anti-inflammatory activity. It is the C-14 acetylation product of Rhodojaponen III, presented as a white to off-white solid. This high-purity compound (99.96%) is intended strictly for research purposes.
  • Diterpenoid with anti-inflammatory activity
  • C-14 acetylation product of Rhodojaponen III
  • Appearance as a white to off-white solid
  • High purity of 99.96%
  • Suitable for research applications
Catalog No. 50-004-18351 Supplier Medchemexpress LLC Supplier No. HYN21545MG
Rhodojaponin V is a diterpenoid isolated from the leaves of *Rhododendron molle* and is recognized for its anti-inflammatory activity. It is the C-14 acetylation product of Rhodojaponen III, presented as a white to off-white solid. This high-purity compound (99.96%) is intended strictly for research purposes.
  • Diterpenoid with anti-inflammatory activity
  • C-14 acetylation product of Rhodojaponen III
  • Appearance as a white to off-white solid
  • High purity of 99.96%
  • Suitable for research applications