Learn More
ATCC Vibrio campbellii; MM32
Catalog No. 502380796
Encompass
Vibrio campbellii strain MM32 is a whole-genome sequenced bacterium that was derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This culture is one of several strains included in a “quorum sensing kit” (ATCC MP-6) developed by the depositor. This strain has the genotype luxN::Cm, luxS::Tn5Kan. It is a mutant sensor strain that is a non-self-stimulating reporter for autoinducer AI-2.
Catalog No. 50-238-0796 Supplier ATCC Supplier No. BAA1121
Vibrio campbellii strain MM32 is a whole-genome sequenced bacterium that was derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This culture is one of several strains included in a “quorum sensing kit” (ATCC MP-6) developed by the depositor. This strain has the genotype luxN::Cm, luxS::Tn5Kan. It is a mutant sensor strain that is a non-self-stimulating reporter for autoinducer AI-2.