Learn More
ATCC Rhodopseudomonas palustris
Catalog No. 502380520
Encompass
Rhodopseudomonas palustris strain CGA009 is a chloramphenicol-resistant derivative of R. palustris strain CGA001. This whole-genome sequenced bacterium can be used in antimicrobial resistance research.
Catalog No. 50-238-0520 Supplier ATCC Supplier No. BAA98
Rhodopseudomonas palustris strain CGA009 is a chloramphenicol-resistant derivative of R. palustris strain CGA001. This whole-genome sequenced bacterium can be used in antimicrobial resistance research.