Learn More
Montana Molecular Rhodopsin P23H BacMam - 5mL
SDP
Supplier:  Montana Molecular N0110N005
Encompass_Preferred
Rhodopsin with a P23H mutation packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. A wild-type version is also available. Co-express with Montana Molecular sensors for ER stress, cAMP, and more. This kit includes the Rhodopsin P23H BacMam and 500mM sodium butyrate in H2O.
Catalog No. NC2600425