Learn More
Montana Molecular Rhodopsin WT BacMam - 5mL
SDP
Supplier:  Montana Molecular N0100N005
Encompass_Preferred
Wild-type Rhodopsin packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. A version with a P23H mutation is also available. Co-express with Montana Molecular sensors for cell stress, cAMP, and more. This kit includes the Wild-type Rhodopsin BacMam and 500mM sodium butyrate in H2O.
Catalog No. NC2600315