Learn More
ATCC Rhodospirillum rubrum
Catalog No. 502385098
Encompass
Rhodospirillum rubrum strain NCIB 8255 is a whole-genome sequenced type strain. This phototroph is tolerant to oxygen, so strictly anoxic conditions are not required when the vial is thawed.
Catalog No. 50-238-5098 Supplier ATCC Supplier No. 11170
Rhodospirillum rubrum strain NCIB 8255 is a whole-genome sequenced type strain. This phototroph is tolerant to oxygen, so strictly anoxic conditions are not required when the vial is thawed.