Learn More
ATCC Vibrio cholerae
Supplier:  ATCC 25870
Encompass
Vibrio cholerae strain 569B is a whole-genome sequenced bacterium that was isolated from a cholera patient. This strain is typed as Inaba serovar O:1 and it is cited to be a good toxin producer.
Catalog No. 50-238-1171