Learn More
Aviva Systems Biology Corp RHOF Antibody - middle region (ARP57310_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57310_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RHOF. Alternate names: ARHF, FLJ20247, RIFApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-62125