Learn More
Aviva Systems Biology Corp RHOG Antibody - C-terminal region (ARP60186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOG. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RHOG. Alternate names: ARHG, MGC125835, MGC125836ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-971