Learn More
TARGETMOL CHEMICALS INC RHOSIN HYDROCHLORIDE 5MG
SDP
Catalog No. 502581159
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rhosin hydrochloride is an effective and specific RhoA subfamily Rho GTPases inhibitor which specifically binds to RhoA to inhibit RhoA-GEF interaction (Kd ~ 0.4 uM). Rhosin hydrochloride does not interact with Cdc42 or Rac1 nor the GEF LARG. Rhosin hydrochloride causes cell apoptosis. purity: 98%
Catalog No. 50-258-1159 Supplier TARGETMOL CHEMICALS INC Supplier No. T167455MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rhosin hydrochloride is an effective and specific RhoA subfamily Rho GTPases inhibitor which specifically binds to RhoA to inhibit RhoA-GEF interaction (Kd ~ 0.4 uM). Rhosin hydrochloride does not interact with Cdc42 or Rac1 nor the GEF LARG. Rhosin hydrochloride causes cell apoptosis. purity: 98%