Learn More
Medchemexpress LLC HY-12646 5mg Medchemexpress, Rhosin (hydrochloride) CAS:1281870-42-5 Purity:>98%
SDP
Catalog No. 502030061
Encompass_Preferred
Medchemexpress, HY-12646 5mg Rhosin (hydrochloride) CAS:1281870-42-5 Rhosin hydrochloride is a potent, specific RhoA subfamily Rho GTPases inhibitor, which specifically binds to RhoA to inhibit RhoA-GEF interaction with a Kd of ~ 0.4 uM, and does not interact with Cdc42 or Rac1, nor the GEF, LARG. Rhosin hydrochloride induces cell apoptosis[2]. Rhosin hydrochloride promotes stress resiliency through enhancing D1-MSN plasticity and reducing hyperexcitability[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0061 Supplier Medchemexpress LLC Supplier No. HY126465MG
Medchemexpress, HY-12646 5mg Rhosin (hydrochloride) CAS:1281870-42-5 Rhosin hydrochloride is a potent, specific RhoA subfamily Rho GTPases inhibitor, which specifically binds to RhoA to inhibit RhoA-GEF interaction with a Kd of ~ 0.4 uM, and does not interact with Cdc42 or Rac1, nor the GEF, LARG. Rhosin hydrochloride induces cell apoptosis[2]. Rhosin hydrochloride promotes stress resiliency through enhancing D1-MSN plasticity and reducing hyperexcitability[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.