Learn More
Selleck Chemical LLC Rhosin hydrochloride S8988-5MG
SDP
Catalog No. 5000025112
Encompass_Preferred
Rhosin (G04) hydrochloride (HCl) is a potent specific inhibitor of RhoA subfamily Rho GTPases with Kd of 0 4 uM Rhosin hydrochloride induces cell apoptosis
Catalog No. 50-000-25112 Supplier Selleck Chemical LLC Supplier No. S89885MG
Rhosin (G04) hydrochloride (HCl) is a potent specific inhibitor of RhoA subfamily Rho GTPases with Kd of 0 4 uM Rhosin hydrochloride induces cell apoptosis