Learn More
Cayman Chemical Rhosin (hydrochloride), Purity: ≥98%, CAS: 1281870-42-5, 10 mg
SDP
Catalog No. 502912528
Encompass_Preferred
An inhibitor of the Rho-GEF protein-protein interaction; inhibits RhoA activity at 10 and 30 µM; selectively reduces the number and size of MCF-7 breast cancer cell mammospheres over MCF-10A non-cancerous cell mammospheres; inhibits migration and invasion of MCF-7 cells and human mammary epithelial cells expressing RhoC; increases neurite outgrowth in PC12 cells at 30 µM; prevents social defeat stress-induced hyperexcitability and increases spine density in D1-MSNs when administered post-social defeat stress in a mouse model of depression at 40 mg/kg; prevents social defeat stress-induced social avoidance and reductions in sucrose preference in a mouse model of depression
Catalog No. 50-291-2528 Supplier Cayman Chemical Supplier No. 3641510 MG
An inhibitor of the Rho-GEF protein-protein interaction; inhibits RhoA activity at 10 and 30 µM; selectively reduces the number and size of MCF-7 breast cancer cell mammospheres over MCF-10A non-cancerous cell mammospheres; inhibits migration and invasion of MCF-7 cells and human mammary epithelial cells expressing RhoC; increases neurite outgrowth in PC12 cells at 30 µM; prevents social defeat stress-induced hyperexcitability and increases spine density in D1-MSNs when administered post-social defeat stress in a mouse model of depression at 40 mg/kg; prevents social defeat stress-induced social avoidance and reductions in sucrose preference in a mouse model of depression