Learn More
Aviva Systems Biology Corp RHOT1 Antibody - N-terminal region (ARP44817_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44817_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOT1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RHOT1. Alternate names: ARHT1, FLJ11040, FLJ12633, MIRO-1, MIRO1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58904