Learn More
Aviva Systems Biology Corp RHOU Antibody - C-terminal region (ARP41274_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41274_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOU. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RHOU. Alternate names: ARHU, CDC42L1, DJ646B12.2, FLJ10616, WRCH1, fJ646B12.2, hG28KApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53292