Learn More
Aviva Systems Biology Corp Rhou Antibody - N-terminal region (ARP41273_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41273_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rhou. It was validated on Western Blot. . Alternate names: 2310026M05Rik, AI182090, Arhu, CDC42L1, G28K, WRCH-1, WRCH1, mG28KApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-284-921