Learn More
Aviva Systems Biology Corp RHOX11 Antibody - N-terminal region (ARP37617_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37617_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOX11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse RHOX11. Alternate names: Gm39, BC049711ApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-56384