Learn More
Aviva Systems Biology Corp RHOXF1 Antibody - C-terminal region (ARP39827_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39827_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOXF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RHOXF1. Alternate names: MGC119030, MGC119033, OTEX, PEPP1ApplicationWB. Species reactivity: Human
Catalog No. 50-284-836