Learn More
Aviva Systems Biology Corp RHOXF2 Antibody - middle region (ARP37789_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37789_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RHOXF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RHOXF2. Alternate names: PEPP-2, PEPP2, THG1, CT107ApplicationIHC, WB. Species reactivity: Human, Pig
Catalog No. 50-014-53094