Learn More
Medchemexpress LLC HY-101089 5mg , RHPS4 CAS:390362-78-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010895MG
Encompass_Preferred
HY-101089 5mg RHPS4 CAS: 390362-78-4 RHPS4 is a potent telomerase inhibitor (IC50 = 0.33 μM). RHPS4 is a DNA damage inducer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1246