Learn More
Akron Biotechnology LLC Recombinant Human Basic Fibroblast Growth Factor-155 Lyophilized, GMP Grade, 100 μg vial, AK9784-0100
Catalog No. NC3136984
Encompass
Akron’s Recombinant Human Basic Fibroblast Growth Factor-155 (rHu bFGF-155) product is manufactured following all relevant cGMP guidelines for ancillary materials.Product Features:Manufacturing•Expressed in E. coli•Animal-derived component free•Tag-free pharmaceutical biosimilar processing•In-house sterile filtration with aseptic filling and lyophilization•All raw materials are compliant, controlled, and traceable under Akron’s Quality Management System (QMS)Stability•Currently on a long-term stability program•Store at 2-8 °C•Transport on cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC3136984 Supplier Akron Biotechnology LLC Supplier No. AK97840100
Akron’s Recombinant Human Basic Fibroblast Growth Factor-155 (rHu bFGF-155) product is manufactured following all relevant cGMP guidelines for ancillary materials.Product Features:Manufacturing•Expressed in E. coli•Animal-derived component free•Tag-free pharmaceutical biosimilar processing•In-house sterile filtration with aseptic filling and lyophilization•All raw materials are compliant, controlled, and traceable under Akron’s Quality Management System (QMS)Stability•Currently on a long-term stability program•Store at 2-8 °C•Transport on cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.