Learn More
Akron Biotechnology LLC Recombinant Human Interleukin-15 Lyophilized, GMP Grade, 40 μg vial, AK9823-0040
Supplier:  Akron Biotechnology LLC AK98230040
Encompass
Akron’s Recombinant Human Interleukin-15 (rHu IL-15) is manufactured following all relevant cGMP guidelines for ancillary materials.Product Features: Active Substance• Carrier protein-free formulation• Animal-free expression system with E. coli host• All raw materials are compliant, controlled, and traceable under Akron’s Quality Management System (QMS)Manufacturing• Type II eCTD MF (#026085) on file with FDA• Tag-free pharmaceutical processing• Gram-scale production capacity• In-house sterile filtration with aseptic filling and lyophilization• No animal-derived materials are used in the manufacture of this productStability• 24-month shelf life• Store at 2-8 °C• Transport with cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications only. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC1955365