Learn More
Akron Biotechnology LLC Recombinant Human Interleukin-7 Lyophilized, GMP Grade, 40 μg vial, AK9842-0040
Supplier:  Akron Biotechnology LLC AK98420040
Encompass
Akron’s Recombinant Human Interleukin-7 (rHu IL-7) is manufactured following all relevant cGMP guidelines for ancillary materials.Active Substance• Carrier protein-free formulation• E. coli expression system• All raw materials are compliant, controlled, and traceable under Akron’s Quality Management System (QMS)Manufacturing• Type II eCTD MF (#026084) on file with FDA• Tag-free pharmaceutical processing• Gram-scale production capacity• In-house sterile filtration with aseptic filling and lyophilizationStability• 24-month shelf life• Store at 2-8 °C• Transport with cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications only. cGMP grade recombinant protein products are not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC1955366