Learn More
Eurofins Discoverx Recombinant Human Activin A, 10 µg. (92-1314)
Supplier:  Eurofins Discoverx 921314
Encompass
Activin A (also known as Inhibin, Beta A) is a protein homodimer of beta A subunits of pituitary FSH (Follicle-Stimulating Hormone) secretion inhibitor. With the beta B subunit, Activin AB stimulates FSH secretion. Activin regulates gonadal stromal cell proliferation negatively and is suspected to have tumor-suppressor activity. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9827