Learn More
Eurofins Discoverx Recombinant Human b-NGF, 20 µg. (92-1023)
Supplier:  Eurofins Discoverx 921023
Encompass
β-NGF is a potent neurotrophic factor that acts as an agonist with high affinity for TrkA, and lower affinity for LNGFR/β-NGFR. It induces the PI 3-kinase, Ras, and PLC signaling pathways, playing a crucial role in the development and preservation of the sensory and sympathetic nervous systems. β-NGF also acts as a growth and differentiation factor for B lymphocytes and enhances pancreatic beta cell survival in the body. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9572