Learn More
Eurofins Discoverx Recombinant Human CCL13, 5 µg. (92-1003)
Supplier:  Eurofins Discoverx 921003
Encompass
CCL13 is a cytokine protein involved in the induction of chemotaxis in monocytes, as well as eosinophils, T lymphocytes, and basophils, by acting as an agonist for cell surface receptors like CCR2, CCR3, and CCR5. It plays a role in leukocyte accumulation during inflammation. Activity of this protein has been linked to asthma and atopic dermatitis, and can be induced by the inflammatory cytokines, including interleukin-1 and TNF-α. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9553