Learn More
Eurofins Discoverx Recombinant Human CCL2, 5 µg. (92-1001)
Supplier:  Eurofins Discoverx 921001
Encompass
CCL2 is a protein involved in chemoattraction which attracts monocytes, activated T cells, basophils, NK cells, and immature dendritic cells to the site of inflammation. CCL2 acts as an agonist on CCR receptors including CCR2, CCR3, and CCR5. Additionally, it is believed to reduce IL-12 production by dendritic cells and promote a Th2 phenotype in CD4+ T cells. Implicated in the pathology of multiple sclerosis, atherosclerosis, and allergic asthma. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9551