Learn More
Eurofins Discoverx Recombinant Human CXCL11, 5 µg. (92-1010)
Catalog No. 501999559
Encompass
CXCL11 is a small cytokine protein that acts as a chemoattractant for IL-2-activated T cells, but does not affect freshly isolated unstimulated T cells, neutrophils, or monocytes. CXCL11 is a "non-ELR" chemokine that is regulated by interferons and elicits its effects by acting as an agonist for the CXCR3 receptor. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9559 Supplier Eurofins Discoverx Supplier No. 921010
CXCL11 is a small cytokine protein that acts as a chemoattractant for IL-2-activated T cells, but does not affect freshly isolated unstimulated T cells, neutrophils, or monocytes. CXCL11 is a "non-ELR" chemokine that is regulated by interferons and elicits its effects by acting as an agonist for the CXCR3 receptor. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.