Learn More
Eurofins Discoverx Recombinant Human CXCL13, 20 µg. (92-1012)
Supplier:  Eurofins Discoverx 921012
Encompass
CXCL13 is a small cytokine protein that is selectively chemotactic for B cells belonging to both the B-1 and B-2 subsets, and carries out its effects by interacting with its chemokine receptor, CXCR5. It takes part in the formation of ectopic lymphoid tissue in chronic inflammation and is expressed highly in the liver, spleen, lymph nodes, and the gut of humans. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9561