Learn More
Eurofins Discoverx Recombinant Human EG-VEGF, 5 µg. (92-1022)
Supplier:  Eurofins Discoverx 921022
Encompass
EG-VEGF (PK1) is a prokinectin growth factor expressed in the ovaries, testis, adrenal, and placental tissues, and is one of the main non-glycosylated proteins in the body. It acts as an angiogenic mitogen and has chemotactic function. EG-VEGF acts as an agonist on PKR1 and PKR2. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9571
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.