Learn More
Eurofins Discoverx Recombinant human EGF, 100 µg. (92-1113)
Supplier:  Eurofins Discoverx 921113
Encompass
Epidermal growth factor (EGF) is a 53 amino acid polypeptide growth factor that binds EGF receptor (EGFR) and stimulates cell growth, proliferation, differentiation, and survival. It is a potent mitogenic factor, and defects result in hypomagnesemia type 4. EGF is an agonist for EGFR. Provided as 100 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9648