Learn More
Eurofins Discoverx Recombinant Human FGF-basic, 10 µg. (92-1027)
Supplier:  Eurofins Discoverx 921027
Encompass
FGF basic (FGF2) belongs to the family of heparin-binding growth factors. It functions as a wide-spectrum mitogenic, angiogenic, and neurotrophic factor and stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal, and endothelial cells. FGF basic has also been implicated in a multitude of physiological and pathological processes, including limb development, angiogenesis, wound healing, and tumor growth. FGF basic is an agonist for FGFR1, FGFR2, FGFR3 and FGFR4. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9575