Learn More
Eurofins Discoverx Recombinant Human GDNF, 2 µg. (92-1299)
Supplier:  Eurofins Discoverx 921299
Encompass
Glial-derived neurotrophic factor (GDNF) is a disulfide-linked, homodimeric neurotrophic factor protein that promotes the survival and differentiation of dopaminergic neurons in culture and is able to prevent apoptosis of motor neurons induced by axotomy. GDNF specifically promotes dopamine uptake and survival, and morphological differentiation of midbrain neurons. It is an agonist for GFRα1. Provided as 2μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9815