Learn More
Eurofins Discoverx Recombinant Human GM-CSF, 5 µg. (92-1317)
Supplier:  Eurofins Discoverx 921317
Encompass
Granulocyte-macrophage colony-stimulating factor (GM-CSF or colony stimulating factor 2 (CSF2)) functions as a cytokine, affects mature cells of the immune system, and plays a role in embryonic development by functioning as an embryokine produced by the reproductive tract. It is an agonist for CSF2RA and CSF2RB. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9830