Learn More
Eurofins Discoverx Recombinant Human IFN-beta, 20 µg. (92-1320)
Supplier:  Eurofins Discoverx 921320
Encompass
IFN-β, a human type I interferon, is produced by fibroblasts and induces non-specific resistance against a broad range of viral infections. It also affects cell proliferation and modulates immune responses. Acts as an agonist for IFNAR1. Provided as 200 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9833