Learn More
Eurofins Discoverx Recombinant Human IGF-I, 100 µg. (92-1030)
Supplier:  Eurofins Discoverx 921030
Encompass
Human IGF-1 is a hormone similar in molecular structure to insulin and plays an important role in childhood growth. Typically exists bound to IGF-BP 1-6, and acts as an agonist for IGF1R. A synthetic analog of IGF-1, mecasermin, is commonly used for the treatment of growth failure. Provided as 100 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9576