Learn More
Eurofins Discoverx Recombinant Human IL-10, 10 µg. (92-1252)
Supplier:  Eurofins Discoverx 921252
Encompass
Interleukin 10 (IL-10) is an anti-inflammatory cytokine protein and mediator of the immune response. Induces pleiotropic effects in immunoregulation and inflammation, enhances B cell survival and proliferation, and induces antibody production. Activates the JAK-STAT signaling pathway, and blocks NFkB activity and synthesis of multiple cytokines, including IFN-gamma, IL-2, IL-3, TNF, and GM-CSF. Agonist for IL10RA and IL10RB. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9773