Learn More
Eurofins Discoverx Recombinant Human IL-15, 10 µg. (92-1265)
Supplier:  Eurofins Discoverx 921265
Encompass
Interleukin 15 (IL-15) is a cytokine protein and mediator of the immune response. Similar to IL-2, IL-15 may compete for the same hematopoietin IL-2 receptors. Agonist for IL15RA. IL-15 is secreted by mononuclear phagocytes in response to viral infection and is responsible for regulation of T-cell and natural killer cell activation and proliferation. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9784