Learn More
Eurofins Discoverx Recombinant Human IL-2, 10 µg. (92-1253)
Supplier:  Eurofins Discoverx 921253
Encompass
Interleukin 2 (IL-2) is a cytokine protein and mediator of the immune response. An agonist for IL2RA and IL2RB, IL-2 regulates activation of T and B lymphocyte proliferation, and also stimulates monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9774